accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10042-59-8

CAS RN 10042-59-8

IUPAC Name: 2-propylheptan-1-ol
Synonyms: 1-heptanol, 2-propyl- propylheptyl alcohol
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: YLQLIQIAXYRMDL-UHFFFAOYNA-N
SMILES: CCCCCC(CO)CCC

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products