accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10049-16-8

CAS RN 10049-16-8

IUPAC Name: vanadium pentafluoride
Synonyms: vanadium pentafluoride
Molecular Weight (g/mol): 145.93
Molecular Formula: F5V
InChi Key: LYTSFXQJUMZNFC-UHFFFAOYSA-N
SMILES: [F].[F].[F].[F].[F].[V]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products