accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 100516-54-9

CAS RN 100516-54-9

IUPAC Name: benzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate
Synonyms: benzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate
Molecular Weight (g/mol): 387.44
Molecular Formula: C24H21NO4
InChi Key: HECRUWTZAMPQOS-UHFFFAOYNA-N
SMILES: O=C(OCC1=CC=CC=C1)N1CC(=O)OC(C1C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products