man

Test

CAS RN 10052-47-8

ClCH3NCH3H3CH3C

CAS RN 10052-47-8

IUPAC Name: triethyl(methyl)azanium chloride
Synonyms: triethyl(methyl)azanium chloride
Molecular Weight (g/mol): 151.68
Molecular Formula: C7H18ClN
InChi Key: NIUZJTWSUGSWJI-UHFFFAOYSA-M
SMILES: [Cl-].CC[N+](C)(CC)CC

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results