accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1007-16-5

CAS RN 1007-16-5

IUPAC Name: 3-bromo-4-fluorobenzoate
Synonyms: 3-bromo-4-fluorobenzoate
Molecular Weight (g/mol): 218
Molecular Formula: C7H3BrFO2
InChi Key: ONELILMJNOWXSA-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(F)C(Br)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products