man

Test

CAS RN 10099-59-9

LaOHNOOOHNOOOHNOO

CAS RN 10099-59-9

IUPAC Name: lanthanum(3+) tris(nitric acid)
Synonyms: lanthanum(3+) tris(nitric acid)
Molecular Weight (g/mol): 327.94
Molecular Formula: H3LaN3O9
InChi Key: PAPXNISCLWTJQS-UHFFFAOYSA-N
SMILES: [La+3].O[N+]([O-])=O.O[N+]([O-])=O.O[N+]([O-])=O
Quantity 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results