accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 100999-26-6

CAS RN 100999-26-6

IUPAC Name: (2R,11R)-10-methyl-10,15-diazapentacyclo[11.6.1.0²,¹¹.0³,⁸.0¹⁶,²⁰]icosa-1(20),3,5,7,13,16,18-heptaene
Synonyms: (2R,11R)-10-methyl-10,15-diazapentacyclo[11.6.1.0²,¹¹.0³,⁸.0¹⁶,²⁰]icosa-1(20),3,5,7,13,16,18-heptaene
Molecular Weight (g/mol): 274.37
Molecular Formula: C19H18N2
InChi Key: WRNKIDLXXXIELU-IEBWSBKVSA-N
SMILES: CN1CC2=CC=CC=C2[C@H]2[C@H]1CC1=CNC3=CC=CC2=C13
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products