man

Test

CAS RN 101-31-5

CH3NOOOH

CAS RN 101-31-5

IUPAC Name: (1R,5S)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl (2S)-3-hydroxy-2-phenylpropanoate
Synonyms: hyoscyamine, its salts and derivatives
Molecular Weight (g/mol): 289.38
Molecular Formula: C17H23NO3
InChi Key: RKUNBYITZUJHSG-VFSICIBPSA-N
SMILES: CN1[C@H]2CC[C@@H]1CC(C2)OC(=O)[C@H](CO)C1=CC=CC=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results