man

Test

CAS RN 101-70-2

H3CONHOCH3

CAS RN 101-70-2

IUPAC Name: 4-methoxy-N-(4-methoxyphenyl)aniline
Synonyms: 4-methoxy-N-(4-methoxyphenyl)aniline
Molecular Weight (g/mol): 229.28
Molecular Formula: C14H15NO2
InChi Key: VCOONNWIINSFBA-UHFFFAOYSA-N
SMILES: COC1=CC=C(NC2=CC=C(OC)C=C2)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results