accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1011531-27-3

CAS RN 1011531-27-3

IUPAC Name: 3-[4-({4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methoxy)phenyl]-3-(1,2-oxazol-3-yl)propanoic acid
Synonyms: 3-[4-({4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazol-5-yl}methoxy)phenyl]-3-(1,2-oxazol-3-yl)propanoic acid
Molecular Weight (g/mol): 488.48
Molecular Formula: C24H19F3N2O4S
InChi Key: BMLGZNVPWRUVNM-UHFFFAOYNA-N
SMILES: CC1=C(COC2=CC=C(C=C2)C(CC(O)=O)C2=NOC=C2)SC(=N1)C1=CC=C(C=C1)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products