man

Test

CAS RN 10127-02-3

HClClClZnH3CNCH3NNCH3CH3

CAS RN 10127-02-3

IUPAC Name: zinc(2+) hydrogen N3,N3,N6,N6-tetramethylacridine-3,6-diamine trichloride
Synonyms: zinc(2+) hydrogen acridine orange trichloride
Molecular Weight (g/mol): 438.1
Molecular Formula: C17H20Cl3N3Zn
InChi Key: VADJQOXWNSPOQA-UHFFFAOYSA-L
SMILES: [H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Color 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)