accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10128-91-3

CAS RN 10128-91-3

IUPAC Name: methyl 2-oxo-1,2-dihydropyridine-3-carboxylate
Synonyms: methyl 2-oxo-1H-pyridine-3-carboxylate
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: SILBTMNCGYLTOK-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CNC1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products