man

Test

CAS RN 1017778-08-3

CH3FFFOOH

CAS RN 1017778-08-3

IUPAC Name: 3-[2-methyl-5-(trifluoromethyl)phenyl]prop-2-enoic acid
Synonyms: 3-[2-methyl-5-(trifluoromethyl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 230.19
Molecular Formula: C11H9F3O2
InChi Key: FJMNLFRMBNXMRQ-UHFFFAOYSA-N
SMILES: CC1=C(C=CC(O)=O)C=C(C=C1)C(F)(F)F

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results