man

Test

CAS RN 1017778-60-7

CH3OOHFF

CAS RN 1017778-60-7

IUPAC Name: 3,4-difluoro-5-methylbenzoic acid
Synonyms: 3,4-difluoro-5-methylbenzoic acid
Molecular Weight (g/mol): 172.13
Molecular Formula: C8H6F2O2
InChi Key: RNAYJQUWVKDTJZ-UHFFFAOYSA-N
SMILES: CC1=C(F)C(F)=CC(=C1)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results