accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10191-60-3

CAS RN 10191-60-3

IUPAC Name: [bis(methylsulfanyl)methylidene](cyano)amine
Synonyms: [bis(methylsulfanyl)methylidene](cyano)amine
Molecular Weight (g/mol): 146.23
Molecular Formula: C4H6N2S2
InChi Key: IULFXBLVJIPESI-UHFFFAOYSA-N
SMILES: CSC(SC)=NC#N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products