man

Test

CAS RN 102-49-8

H2NClCl

CAS RN 102-49-8

IUPAC Name: 1-(3,4-dichlorophenyl)methanamine
Synonyms: 1-(3,4-dichlorophenyl)methanamine
Molecular Weight (g/mol): 176.04
Molecular Formula: C7H7Cl2N
InChi Key: IXHNFOOSLAWRBQ-UHFFFAOYSA-N
SMILES: NCC1=CC=C(Cl)C(Cl)=C1
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)