accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 102-84-1

CAS RN 102-84-1

IUPAC Name: tris(prop-2-en-1-yl) phosphite
Synonyms: tris(prop-2-en-1-yl) phosphite
Molecular Weight (g/mol): 202.19
Molecular Formula: C9H15O3P
InChi Key: KJWHEZXBZQXVSA-UHFFFAOYSA-N
SMILES: C=CCOP(OCC=C)OCC=C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products