CAS RN 1021868-77-8
CAS RN 1021868-77-8
IUPAC Name:
tetrasodium 3-(2-{4-formyl-6-methyl-5-oxo-3-[(phosphonatooxy)methyl]-2,5-dihydropyridin-2-ylidene}hydrazin-1-yl)-7-nitronaphthalene-1,5-disulfonate
Synonyms:
tetrasodium 3-(2-{4-formyl-6-methyl-5-oxo-3-[(phosphonatooxy)methyl]pyridin-2-ylidene}hydrazin-1-yl)-7-nitronaphthalene-1,5-disulfonate
Molecular Weight (g/mol):
694.35
Molecular Formula:
C18H11N4Na4O14PS2
InChi Key:
TXFLDKHOZFWVAC-UHFFFAOYSA-J
SMILES:
[Na+].[Na+].[Na+].[Na+].CC1=NC(=NNC2=CC(=C3C=C(C=C(C3=C2)S([O-])(=O)=O)[N+]([O-])=O)S([O-])(=O)=O)C(COP([O-])([O-])=O)=C(C=O)C1=O
1
–
1
of
1
results
1
PPNDS, Tocris Bioscience™
CAS: 1021868-77-8 Molecular Formula: C18H11N4Na4O14PS2 Molecular Weight (g/mol): 694.353 InChI Key: TXFLDKHOZFWVAC-UHFFFAOYSA-J Synonym: ppnds tetrasodium PubChem CID: 57369378 IUPAC Name: tetrasodium;3-[2-[4-formyl-6-methyl-5-oxo-3-(phosphonatooxymethyl)pyridin-2-ylidene]hydrazinyl]-7-nitronaphthalene-1,5-disulfonate SMILES: CC1=NC(=NNC2=CC(=C3C=C(C=C(C3=C2)S(=O)(=O)[O-])[N+](=O)[O-])S(=O)(=O)[O-])C(=C(C1=O)C=O)COP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]