CAS RN 10231-84-2
CAS RN 10231-84-2
IUPAC Name:
(2S,3S,4R,5S,6S)-2-methyl-6-(4-nitrophenoxy)oxane-3,4,5-triol
Synonyms:
(2S,3S,4R,5S,6S)-2-methyl-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular Weight (g/mol):
285.25
Molecular Formula:
C12H15NO7
InChi Key:
YILIDCGSXCGACV-SQKFTNEHSA-N
SMILES:
C[C@@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@@H](O)[C@H](O)[C@@H]1O