man

Test

CAS RN 10236-60-9

OOClCl

CAS RN 10236-60-9

IUPAC Name: 2-(2,3-dichlorophenyl)acetate
Synonyms: 2-(2,3-dichlorophenyl)acetate
Molecular Weight (g/mol): 204.03
Molecular Formula: C8H5Cl2O2
InChi Key: YWMXEUIQZOQESD-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=CC=CC(Cl)=C1Cl
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results