man

Test

CAS RN 1024-57-3

ClOClClClClClCl

CAS RN 1024-57-3

IUPAC Name: 1,6,8,9,10,11,11-heptachloro-4-oxatetracyclo[6.2.1.0²,⁷.0³,⁵]undec-9-ene
Synonyms: heptachlor epoxide C10H5cl7O heptachlor-epoxide HCE epoxyheptachlor heptachlorepoxide (substance) epoxide, heptachlor velsicol 53-CS-17 heptachlorepoxide
Molecular Weight (g/mol): 389.3
Molecular Formula: C10H5Cl7O
InChi Key: ZXFXBSWRVIQKOD-UHFFFAOYNA-N
SMILES: ClC1C2OC2C2C1C1(Cl)C(Cl)=C(Cl)C2(Cl)C1(Cl)Cl
Quantity 
  • (4)
  • (2)
For Use With (Equipment) 
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (2)