accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10242-11-2

CAS RN 10242-11-2

IUPAC Name: 5-bromo-1-benzofuran-2-carboxylic acid
Synonyms: 5-bromo-1-benzofuran-2-carboxylic acid
Molecular Weight (g/mol): 241.04
Molecular Formula: C9H5BrO3
InChi Key: QKUWZCOVKRUXKX-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=CC(Br)=CC=C2O1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products