CAS RN 10270-29-8
CAS RN 10270-29-8
IUPAC Name:
4-pentylbenzonitrile
Synonyms:
4-pentylbenzonitrile
Molecular Weight (g/mol):
173.26
Molecular Formula:
C12H15N
InChi Key:
LGPQFJXPPKNHJU-UHFFFAOYSA-N
SMILES:
CCCCCC1=CC=C(C=C1)C#N
1
–
1
of
1
results
1
4-Pentylbenzonitrile, 97%, Thermo Scientific™
CAS: 10270-29-8 Molecular Formula: C12H15N Molecular Weight (g/mol): 173.26 MDL Number: MFCD00218832 InChI Key: LGPQFJXPPKNHJU-UHFFFAOYSA-N Synonym: 4-n-pentylbenzonitrile,4-pentylbenzenecarbonitrile,p-amylbenzonitrile,p-n-pentylbenzonitrile,benzonitrile, 4-pentyl PubChem CID: 264951 IUPAC Name: 4-pentylbenzonitrile SMILES: CCCCCC1=CC=C(C=C1)C#N