accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 103-81-1

CAS RN 103-81-1

IUPAC Name: 2-phenylacetamide
Synonyms: phenylacetamide phenyl-β-acetylamine α-toluamide
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: LSBDFXRDZJMBSC-UHFFFAOYSA-N
SMILES: NC(=O)CC1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products