accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 103011-38-7

CAS RN 103011-38-7

IUPAC Name: 3-(1,3-dioxolan-2-yl)thiophene-2-sulfonyl chloride
Synonyms: 3-(1,3-dioxolan-2-yl)thiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 254.7
Molecular Formula: C7H7ClO4S2
InChi Key: RRTOURREASJQSR-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=C(C=CS1)C1OCCO1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products