CAS RN 103068-41-3
CAS RN 103068-41-3
IUPAC Name:
4-fluorobenzene-1,3-diol
Synonyms:
4-fluorobenzene-1,3-diol
Molecular Weight (g/mol):
128.1
Molecular Formula:
C6H5FO2
InChi Key:
XPOIJNIQXJYQOV-UHFFFAOYSA-N
SMILES:
OC1=CC(O)=C(F)C=C1