accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10312-55-7

CAS RN 10312-55-7

IUPAC Name: 2-aminobenzene-1,4-dicarboxylate
Synonyms: 2-aminobenzene-1,4-dicarboxylate
Molecular Weight (g/mol): 179.13
Molecular Formula: C8H5NO4
InChi Key: GPNNOCMCNFXRAO-UHFFFAOYSA-L
SMILES: NC1=CC(=CC=C1C([O-])=O)C([O-])=O
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products