man

Test

CAS RN 1031843-28-3

CH3ONNHCH3

CAS RN 1031843-28-3

IUPAC Name: methyl[(4-methyl-2-phenyl-1,3-oxazol-5-yl)methyl]amine
Synonyms: methyl[(4-methyl-2-phenyl-1,3-oxazol-5-yl)methyl]amine
Molecular Weight (g/mol): 202.26
Molecular Formula: C12H14N2O
InChi Key: XCBYKJIAOCVNGY-UHFFFAOYSA-N
SMILES: CNCC1=C(C)N=C(O1)C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results