accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 103360-04-9

CAS RN 103360-04-9

IUPAC Name: (4-fluorophenyl)methanesulfonyl chloride
Synonyms: (4-fluorophenyl)methanesulfonyl chloride
Molecular Weight (g/mol): 208.63
Molecular Formula: C7H6ClFO2S
InChi Key: UUQGWVIRPCRTSA-UHFFFAOYSA-N
SMILES: FC1=CC=C(CS(Cl)(=O)=O)C=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products