CAS RN 10361-37-2
CAS RN 10361-37-2
Barium chloride, 10% w/v aq. soln., Thermo Scientific Chemicals
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L Synonym: barium chloride,barium dichloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508 PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+);dichloride SMILES: [Cl-].[Cl-].[Ba++]
Promotions are available
Barium chloride, anhydrous, 99.998% (metals basis), Thermo Scientific Chemicals
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L Synonym: barium chloride,barium dichloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508 PubChem CID: 25204 ChEBI: CHEBI:63317 SMILES: [Cl-].[Cl-].[Ba++]
Barium chloride, 0.1N Standardized Solution, Thermo Scientific Chemicals
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L Synonym: barium chloride,barium dichloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508 PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+);dichloride SMILES: [Cl-].[Cl-].[Ba++]
Promotions are available
Barium chloride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L Synonym: barium chloride,barium dichloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508 PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+);dichloride SMILES: [Cl-].[Cl-].[Ba++]
Barium chloride, 0.5N Standardized Solution, Thermo Scientific Chemicals
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L Synonym: barium chloride,barium dichloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508 PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+);dichloride SMILES: [Cl-].[Cl-].[Ba++]
Promotions are available
Barium chloride, ultra dry, 99.95% (metals basis), Thermo Scientific™
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L Synonym: barium chloride,barium dichloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508 PubChem CID: 25204 ChEBI: CHEBI:63317 SMILES: [Cl-].[Cl-].[Ba++]
Barium chloride concentrate, For 1L standard solution, 0.05 M BaCl2 (0.1N), Honeywell Fluka™
CAS: 10361-37-2 Molecular Formula: BaCl2 Molecular Weight (g/mol): 208.23 MDL Number: MFCD00003445 InChI Key: WDIHJSXYQDMJHN-UHFFFAOYSA-L Synonym: barium chloride,barium dichloride,barium chloride solution,ccris 2286,barium chloride, anhydrous,barium chloride, ultra dry,bariumchlorid,dichlorobarium,dsstox_cid_24508 PubChem CID: 25204 ChEBI: CHEBI:63317 IUPAC Name: barium(2+);dichloride SMILES: [Cl-].[Cl-].[Ba++]