man

Test

CAS RN 103628-48-4

CH3NHSOOHNNH3CCH3HOOOHO

CAS RN 103628-48-4

IUPAC Name: 1-{3-[2-(dimethylamino)ethyl]-1H-indol-5-yl}-N-methylmethanesulfonamide; butanedioic acid
Synonyms: succinic acid; sumatriptan
Molecular Weight (g/mol): 413.49
Molecular Formula: C18H27N3O6S
InChi Key: PORMUFZNYQJOEI-UHFFFAOYSA-N
SMILES: OC(=O)CCC(O)=O.CNS(=O)(=O)CC1=CC=C2NC=C(CCN(C)C)C2=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results