accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10377-58-9

CAS RN 10377-58-9

IUPAC Name: magnesium(2+) diiodide
Synonyms: magnesium iodide
Molecular Weight (g/mol): 278.11
Molecular Formula: I2Mg
InChi Key: BLQJIBCZHWBKSL-UHFFFAOYSA-L
SMILES: [Mg++].[I-].[I-]
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products