man

Test

CAS RN 10377-66-9

MnONOOONOO

CAS RN 10377-66-9

IUPAC Name: manganese(2+) dinitrate
Synonyms: manganese(II) nitrate
Molecular Weight (g/mol): 178.95
Molecular Formula: MnN2O6
InChi Key: MIVBAHRSNUNMPP-UHFFFAOYSA-N
SMILES: [Mn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results