accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10378-49-1

CAS RN 10378-49-1

IUPAC Name: sodium tetrabromogolduide hydrate
Synonyms: sodium tetrabromoaurate(III) hydrate
Molecular Weight (g/mol): 557.59
Molecular Formula: AuBr4H2NaO
InChi Key: DCARRTFPXWVHEC-UHFFFAOYSA-J
SMILES: O.[Na+].Br[Au-](Br)(Br)Br
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products