accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 103878-83-7

CAS RN 103878-83-7

IUPAC Name: hydrogen N-(2-aminoethyl)-5-chloropyridine-2-carboxamide chloride
Synonyms: hydrogen lazabemide chloride
Molecular Weight (g/mol): 236.1
Molecular Formula: C8H11Cl2N3O
InChi Key: JMFKTFLARGGXCC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCNC(=O)C1=CC=C(Cl)C=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products