CAS RN 103980-44-5
CAS RN 103980-44-5
IUPAC Name:
(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-[(furan-2-carbonylsulfanyl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrochloride
Synonyms:
ceftiofur hydrochloride
Molecular Weight (g/mol):
560.01
Molecular Formula:
C19H18ClN5O7S3
InChi Key:
KEQFDTJEEQKVLM-JUODUXDSSA-N
SMILES:
Cl.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC(=O)C3=CC=CO3)=C(N2C1=O)C(O)=O)C1=CSC(N)=N1
1
–
2
of
2
results
1
Thermo Scientific Chemicals Ceftiofur hydrochloride
CAS: 103980-44-5 Molecular Formula: C19H18ClN5O7S3 Molecular Weight (g/mol): 560.01 InChI Key: KEQFDTJEEQKVLM-JUODUXDSSA-N IUPAC Name: (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-[(furan-2-carbonylsulfanyl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid hydrochloride SMILES: Cl.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC(=O)C3=CC=CO3)=C(N2C1=O)C(O)=O)C1=CSC(N)=N1