man

Test

CAS RN 104-76-7

H3CCH3OH

CAS RN 104-76-7

IUPAC Name: 2-ethylhexan-1-ol
Synonyms: 2-ethylhexanol ethylhexanol 2-aethylhexanol
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: YIWUKEYIRIRTPP-UHFFFAOYNA-N
SMILES: CCCCC(CC)CO
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (5)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)