accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 104-96-1

CAS RN 104-96-1

IUPAC Name: 4-(methylsulfanyl)aniline
Synonyms: benzenamine, 4-(methylthio)-
Molecular Weight (g/mol): 139.22
Molecular Formula: C7H9NS
InChi Key: YKFROQCFVXOUPW-UHFFFAOYSA-N
SMILES: CSC1=CC=C(N)C=C1
Boiling Point 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products