accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 104040-78-0

CAS RN 104040-78-0

IUPAC Name: 3-(4,6-dimethoxypyrimidin-2-yl)-1-{[3-(trifluoromethyl)pyridin-2-yl]sulfonyl}urea
Synonyms: flazasulfuron
Molecular Weight (g/mol): 407.32
Molecular Formula: C13H12F3N5O5S
InChi Key: HWATZEJQIXKWQS-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=NC(NC(=O)NS(=O)(=O)C2=NC=CC=C2C(F)(F)F)=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products