accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 104104-50-9

CAS RN 104104-50-9

IUPAC Name: hydrogen 4-[6-imino-3-(4-methoxyphenyl)-1,6-dihydropyridazin-1-yl]butanoic acid bromide
Synonyms: gabexate
Molecular Weight (g/mol): 368.23
Molecular Formula: C15H18BrN3O3
InChi Key: GFZHNFOGCMEYTA-UHFFFAOYSA-N
SMILES: [H+].[Br-].COC1=CC=C(C=C1)C1=NN(CCCC(O)=O)C(=N)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products