man

Test

CAS RN 10424-65-4

H2OH2OH2OH2OH2OOHCH3NCH3CH3H3C

CAS RN 10424-65-4

IUPAC Name: tetramethylazanium pentahydrate hydroxide
Synonyms: tetramethylammonium hydroxide pentahydrate
Molecular Weight (g/mol): 181.23
Molecular Formula: C4H23NO6
InChi Key: MYXKPFMQWULLOH-UHFFFAOYSA-M
SMILES: O.O.O.O.O.[OH-].C[N+](C)(C)C
Percent Purity 
  • (1)
  • (3)
  • (5)
Quantity 
  • (3)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (4)
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (2)
  • (7)
  • (2)
  • (1)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (2)
  • (4)
  • (3)