CAS RN 104322-63-6
CAS RN 104322-63-6
IUPAC Name:
(1R,6S,8S)-11,11-dimethyl-5-oxa-3λ⁶-thia-4-azatetracyclo[6.2.1.0¹,⁶.0⁴,⁶]undecane-3,3-dione
Synonyms:
(1R,6S,8S)-11,11-dimethyl-5-oxa-3λ⁶-thia-4-azatetracyclo[6.2.1.0¹,⁶.0⁴,⁶]undecane-3,3-dione
Molecular Weight (g/mol):
229.29
Molecular Formula:
C10H15NO3S
InChi Key:
GBBJBUGPGFNISJ-KAHWVJTKSA-N
SMILES:
CC1(C)[C@H]2CC[C@@]11CS(=O)(=O)N3O[C@@]13C2
1
–
1
of
1
results
1