CAS RN 10450-59-6
CAS RN 10450-59-6
IUPAC Name:
dicerium(3+) octahydrate trisulfate
Synonyms:
dicerium(3+) octahydrate trisulfate
Molecular Weight (g/mol):
712.52
Molecular Formula:
Ce2H16O20S3
InChi Key:
PQBKXYUMEMUVIH-UHFFFAOYSA-H
SMILES:
O.O.O.O.O.O.O.O.[Ce+3].[Ce+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
1
–
2
of
2
results
1
Cerium(III) sulfate octahydrate, 99.99%, (trace metal basis), Thermo Scientific Chemicals
CAS: 10450-59-6 Molecular Formula: Ce2O12S3·8H2O Molecular Weight (g/mol): 712.55 MDL Number: MFCD00149641
2
Cerium(III) sulfate octahydrate, REacton™, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 10450-59-6 Molecular Formula: Ce2(SO4)3·8H2O MDL Number: MFCD00149641