man

Test

CAS RN 10463-37-3

OHOOHNOONOO

CAS RN 10463-37-3

IUPAC Name: 2-(4-hydroxy-3,5-dinitrophenyl)acetic acid
Synonyms: (4-hydroxy-3,5-dinitrophenyl)acetic acid
Molecular Weight (g/mol): 242.14
Molecular Formula: C8H6N2O7
InChi Key: MLVYQQLUGFSXQH-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(=C(O)C(=C1)[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results