CAS RN 10469-09-7
CAS RN 10469-09-7
IUPAC Name:
tetrachloropyridine-2-carboxylic acid
Synonyms:
tetrachloropyridine-2-carboxylic acid
Molecular Weight (g/mol):
260.88
Molecular Formula:
C6HCl4NO2
InChi Key:
GXFRQLQUKBSYQL-UHFFFAOYSA-N
SMILES:
OC(=O)C1=NC(Cl)=C(Cl)C(Cl)=C1Cl
1
–
1
of
1
results
1
3,4,5,6-Tetrachloropyridine-2-carboxylic acid, 98%, Thermo Scientific Chemicals
CAS: 10469-09-7 Molecular Formula: C6HCl4NO2 Molecular Weight (g/mol): 260.88 MDL Number: MFCD00185833 InChI Key: GXFRQLQUKBSYQL-UHFFFAOYSA-N Synonym: 3,4,5,6-tetrachloropicolinic acid,tetrachloropicolinic acid,tetrachloropyridine-2-carboxylic acid,2-pyridinecarboxylic acid, 3,4,5,6-tetrachloro,unii-7e9zn8ab09,3,4,5,6-tetrachloro-2-pyridinecarboxylic acid,2-pyridinecarboxylicacid, 3,4,5,6-tetrachloro,tetrachloropyridine-2-carboxylicacid,dsstox_cid_24509,dsstox_rid_80279 PubChem CID: 82646 IUPAC Name: 3,4,5,6-tetrachloropyridine-2-carboxylic acid SMILES: OC(=O)C1=NC(Cl)=C(Cl)C(Cl)=C1Cl