accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 104719-63-3

CAS RN 104719-63-3

IUPAC Name: 5-(tert-butoxy)-2-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoic acid
Synonyms: 5-(tert-butoxy)-2-[(tert-butoxycarbonyl)amino]-5-oxopentanoic acid
Molecular Weight (g/mol): 303.36
Molecular Formula: C14H25NO6
InChi Key: YGSRAYJBEREVRB-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CCC(NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products