accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10477-47-1

CAS RN 10477-47-1

IUPAC Name: prop-2-yn-1-yl prop-2-enoate
Synonyms: prop-2-yn-1-yl prop-2-enoate
Molecular Weight (g/mol): 110.11
Molecular Formula: C6H6O2
InChi Key: WPBNLDNIZUGLJL-UHFFFAOYSA-N
SMILES: C=CC(=O)OCC#C
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products
1x1 image pixel for data collection