man

Test

CAS RN 10486-19-8

H3CO

CAS RN 10486-19-8

IUPAC Name: tridecanal
Synonyms: tridecanal
Molecular Weight (g/mol): 198.35
Molecular Formula: C13H26O
InChi Key: BGEHHAVMRVXCGR-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results