accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 105-50-0

CAS RN 105-50-0

IUPAC Name: 1,5-diethyl 3-oxopentanedioate
Synonyms: 1,5-diethyl 3-oxopentanedioate
Molecular Weight (g/mol): 202.21
Molecular Formula: C9H14O5
InChi Key: ZSANYRMTSBBUCA-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)CC(=O)OCC
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products