man

Test

CAS RN 105-58-8

H3COOOCH3

CAS RN 105-58-8

IUPAC Name: diethyl carbonate
Synonyms: diethyl carbonate
Molecular Weight (g/mol): 118.13
Molecular Formula: C5H10O3
InChi Key: OIFBSDVPJOWBCH-UHFFFAOYSA-N
SMILES: CCOC(=O)OCC
Boiling Point 
  • (3)
  • (7)
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (3)
  • (7)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (8)